Drying Cabinet with Glass Doors
Drying Cabinet with Glass Doors
Actual product may vary
Fisherbrand
Drying Cabinet with Glass Doors
Ideal for drying laboratory glassware, surgical instruments, warming, or simply for storage. LEEC™ Drying Cabinet with Glass Doors employs natural convection for drying.
Have Questions?
Catalog NumberCapacity (Metric)Dimensions (D x W x H) InteriorElectrical RequirementsNo. of Shelves
FKS12989968540 L590 x 630 x 1430 mm230 V 50 Hz 1500 w4
Catalog number FKS12989968
Price (SGD)
10,495.00
PK
Add to cart
Capacity (Metric):
540 L
Dimensions (D x W x H) Interior:
590 x 630 x 1430 mm
Electrical Requirements:
230 V 50 Hz 1500 w
No. of Shelves:
4
Price (SGD)
10,495.00
PK
Add to cart
  • Rust-resistant
  • Finished in white powder-coated paint
  • Heatproof, toughened glass doors
  • Adjustable wire mesh shelves
  • Waterproof heating elements
  • Full-length, stainless steel hinges
  • Spring loaded catches
  • Stainless steel sliding shutter for ventilation.
  • Adjustable front feet
Specifications
Capacity (Metric)540 L
Temperature (Metric)50°C
TypeDrying Cabinet
For Use With (Application)Cabinets for drying drained glassware and instruments, or for applications requiring controlled warming
MaterialStainless Steel, Polyester
ModelF2
No. of Shelves4
Dimensions (D x W x H) Interior590 x 630 x 1430 mm
Dimensions (L x W x H)610 x 635 x 1626 mm (External DxWxH)
Width (Metric)635 mm
Height (Metric)1626 mm
Depth (Metric)610 mm
Depth (Metric) Interior590 mm
Weight (Metric)75 kg
Electrical Requirements230 V 50 Hz 1500 w
Voltage230 V
Wattage1500 w
Frequency50 Hz
Unit SizePK