Quickfit™ Reduction Socket Adapter
Quickfit™ Reduction Socket Adapter
Actual product may vary
Fisherbrand
Quickfit™ Reduction Socket Adapter
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCone SizeStandard Taper Joint No.
FKS12989253Cone Size: 34/3524/29
FKS12654891Cone Size: 29/3224/29
Catalog number FKS12989253
Price (SGD)
191.00
PK
Add to cart
Cone Size:
Cone Size: 34/35
Standard Taper Joint No.:
24/29
Price (SGD)
191.00
PK
Add to cart
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
  • Useful for the connection of dissimilar sized ground glass joints
  • Small sized socket at the top, large sized cone at the bottom
Specifications
Cone SizeCone Size: 34/35
For Use With (Equipment)Dissimilar sized ground Glass joint
IncludesSmall socket, large cone
MaterialBorosilicate Glass
Product TypeReduction adapter
Standard Taper Joint No.24/29
Unit SizePK