Wire Gauze with Ceramic Center
Product Image
Fisherbrand
Wire Gauze with Ceramic Center
Provides excellent support to beakers and other glassware during heating on ring stands. R&L Enterprises Wire Gauze with Ceramic Center is a flat square manufactured from galvanized iron mesh.
Have Questions?
Catalog number FKS12988950
Price (SGD)
119.00
PK
Add to cart
Price (SGD)
119.00
PK
Add to cart
  • Non-fraying edges
  • Durable and long-lasting
  • Ceramic center disperses heat evenly
Specifications
Ceramic CenterCeramic Center
Dimensions (L x W)150 x 150 mm
EdgesNon-Fraying
IncludesCeramic center
Quantity10 Pack
ShapeSquare
Length (Metric)150 mm
MaterialIron
TypeWire Gauze
Width (Metric)150 mm
Unit SizePK