Polyurethane Foam Stoppers
Actual product may vary
Fisherbrand
Polyurethane Foam Stoppers
Formed by polymerization in the presence of a foaming agent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS1298429140 mm
FKS1297429135 mm
Catalog number FKS12984291
Price (SGD)
59.00
Pack
Diameter (Metric):
40 mm
Price (SGD)
59.00
Pack
  • Made up of polyurethane
  • Rigid and flexible
Specifications
Diameter (Metric)40 mm
Height (Metric)50 mm
Quantity100/Pk.
MaterialPolyurethane
Product TypePlug Stopper
Unit SizePack