Designed for use in public access areas such as A&E and Out-patients. Sharpsafe™ Polypropylene Medicinal Sharps Container allows easy disposal of sharps but provides added security for both the user and the general public. The Protected Access range also discourages the use of the sharps container for non sharps waste, avoiding unnecessary waste disposal costs.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS12983525
4 L
FKS12903535
7 L
FKS12943525
1.0 L
FKS12963525
2.0 L
4 Options
Catalog number FKS12983525
Price (SGD)
9.00
PK
Add to cart
Capacity (Metric):
4 L
Price (SGD)
9.00
PK
Add to cart
Easy disposal of sharps
Protected Access range
Polypropylene
Specifications
Capacity (Metric)4 L
Certifications/ComplianceBS 7320, AFNOR NX30-500, UN 3291
Closure TypeLid
Closure ColorYellow
ColorYellow
Model4100
Quantity1 Ea.
IncludesClosure
For Use With (Application)Designed for use in public access areas such as A&E and out-patients