pH 700, CON 700, pH-CON 700 pH/Conductivity Benchtop Meters, Measures conductivity, TDS and temperature
pH 700, CON 700, pH-CON 700 pH/Conductivity Benchtop Meters, Measures conductivity, TDS and temperature
Actual product may vary
Fisherbrand
pH 700, CON 700, pH-CON 700 pH/Conductivity Benchtop Meters, Measures conductivity, TDS and temperature
An affordable way to take consistent, accurate conductivity measurements in your laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
FKS12983517PC 700 Meter with pH Electrode
FIS13-620-464PC 700 Meter with pH Electrode
Catalog number FKS12983517
Price (SGD)
4,598.00
EA
Add to cart
Model:
PC 700 Meter with pH Electrode
Price (SGD)
4,598.00
EA
Add to cart
Compact, space-saving design with a larger, easier-to-read display
  • Low cost plus high performance
  • Large dual display
  • Built-in memory function
  • Hold function
  • Built-in memory function
  • Available with built-in electrode stand
  • Selectable manual or Automatic Temperature Compensation
Note: Cat. No. 13-620-461 does not come with a probe.
Specifications
Accuracy (mV)±0.2 within 199.9mV, ±2 beyond 200mV
Accuracy (pH)±0.01
Accuracy (TDS)±1% FS
Calibration Points5
Display TypeLCD
Measurement ModespH, Conductivity, Temperature
Range (Conductivity)0 μS/cm to 200 mS/cm
Range (mV)±2000 mV
Range (pH)0 to 14
Range (TDS)0ppm to 100ppt at 0.5 factor (200ppt at 1 factor)
Resolution (pH)0.01
Temperature Range (Metric)0°C to 100°C
For Use With (Application)Measures pH, mV, conductivity, total dissolved solids (TDS), and temperature (C or F)
IncludesPC 700 meter with pH electrode, conductivity/temperature probe, electrode stand
ModelPC 700 Meter with pH Electrode
Probe TypeConductivity/Temperature
Product TypepH/Con 700
Resolution (Conductivity)0.01μS/cm, 0.1μS/cm, 1μS/cm, 0.01mS/cm, 0.1mS/cm
Resolution (mV)0.1 Within 199.9mV, ±1 beyond 199.9mV
Resolution (TDS)0.01ppm, 0.1ppm, 1ppm, 0.01ppt, 0.1ppt
Temperature CompensationAutomatic or manual from 0° to 100°C
Temperature Range (English)32°F to 212°F
Temperature Resolution0.1°C
Warranty3 Years
Voltage120/240 V
Unit SizeEA