SP Scienceware™ Nessler Tube Racks
SP Scienceware™ Nessler Tube Racks
Actual product may vary
Fisher Catalogue
SP Scienceware™ Nessler Tube Racks
Feature six places, 27mm diameter to hold 50mL tubes
Have Questions?
Catalog NumberPositions
FKS129825506
Catalog number FKS12982550
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Positions:
6
  • Top to bottom deck height is 10cm (4 in.)
  • Flat bottom deck provides a reflective surface for tubes
  • Easy snap together assembly
  • Steam autoclavable
Specifications
Product TypeNessler Tube Rack
AutoclavableNot Autoclavable
HoldsTubes (5 mL)
MaterialPolypropylene
Positions6
Dimensions (L x W x H)11.75 x 3.75 x 5.5 in. (30 x 9.5 x 14cm)
Length (Metric)30 cm
Length (English)11.75 in.
Width (Metric)9.5 cm
Width (English)3.75 in.
Height (Metric)14 cm
Height (English)5.5 in.
Unit SizeEA