Cryobox
Actual product may vary
Fisherbrand
Cryobox
Robust hinge with safe snap-on lid, stackable
Have Questions?
Catalog NumberCapacity (Metric)
FKS129821415 mL
Catalog number FKS12982141
Price (SGD)
124.00
Pack
Capacity (Metric):
5 mL
Price (SGD)
124.00
Pack
Autoclavable, temperature resistant from -90°C to +121°C
  • High quality polypropylene
  • Available in natural and various colours
  • 81 places, 9 x 9 grid, numerically coded
  • Robust hinge with safe snap-on lid
  • Stackable for safe transportation
  • Drain bores at the bottom for dew liquids
  • Removable grid inserts
  • Can also be used for larger vessels by removing the grid inserts
For sample storage and transportation.
Specifications
Capacity (Metric)5 mL
Closure TypeSnap-On Lid
MaterialPolypropylene
ShapeSquare
TypeCryo Box
Holds3 to 5 mL
Max. Temperature (Metric)121°C
Min. Temperature (Metric)-90°C
Quantity5 Pack
Weight (Metric)1 kg
Unit SizePack