Search Thermo Fisher Scientific
Catalog Number | Length (Metric) | Blade Style | Tip Style |
---|---|---|---|
FKS12982055 | 115 mm | Curved | Very Fine Point |
FKS12358099 | 125 mm | Straight | 1 Pointed, 1 Blunt |
FKS12348099 | 115 mm | Straight | Very Fine Point |
FKS12912055 | 125 mm | Straight | Fine Point |
FKS12972055 | 125 mm | Straight | Very Fine Point |
FKS12494070 | 150 mm | Straight | Fine Point |
FKS12932055 | 200 mm | Straight | Very Fine Point |