Eutech™ CyberScan PCD 650 Multi-parameter Meter
Eutech™ CyberScan PCD 650 Multi-parameter Meter
Actual product may vary
Fisherbrand
Eutech™ CyberScan PCD 650 Multi-parameter Meter
Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.
Have Questions?
Catalog NumberRange (pH)
FKS12981758-2 to 20
Catalog number FKS12981758
Price (SGD)
7,896.00
PK
Add to cart
Range (pH):
-2 to 20
Price (SGD)
7,896.00
PK
Add to cart
Eutech™ CyberScan PCD 650 Multi-parameter Meter that measures pH, mV, ion, conductivity, tds, resistivity, salinity and dissolved oxygen. Combining the strengths of all CyberScan 600 series meters, the rugged CyberScan PCD650 is our top-of-the-line waterproof handheld that enables concurrent measurement of pH, conductivity, dissolved oxygen and temperature, all at the same time. Features a One-Glance-Tells-All Screen Display with Backlight Electrode status report, calibration data and simultaneous display of up to 4 parameters.
Features:
  • Simultaneously measures and displays four parameters
  • ±1% full scale accuracy at 3-decimal resolution across a wide measurement range
  • Ion-measurement capabilities with 3-digit resolution
  • Accepts two-cell and four-cell conductivity probe to measure up to 500 m/s — meter measures pure water!
  • Selectable multi-point calibration for more accurate conductivity measurements or single-point calibration for quick, convenient calibration with no mistakes
  • Measures 90.00mg/L in DO concentration and 600% in DO saturation
  • Fast and easy DO calibration with 100% calibration option against atmospheric air or 0% calibration against a zero solution
  • Salinity, Temperature and Pressure Compensation—accurate readings under varying conditions
  • No warm up time required with galvanic system of measurement
  • Easy-to-understand prompt messages for easy set-up, calibration and trouble-shooting
  • Recorded readings individually stamped with time and date in GLP compliant format
  • Auto-logging meter saves readings automatically within seconds to prevent data loss
  • Wireless Information Technology: No wires, no cables—Transfer data from meter to computer with the press of a button
  • Non-Volatile Memory protects information and meter settings, even when batteries run out
  • Four Levels of Password Protection prevents tampering or accidental data deletion
Specifications
OutputsIrDA Wireless Communications
Accuracy (Barometer)0.1 mmHg or 0.1 kPa
Accuracy (Conductivity)±1% FS + 1 LSD
Accuracy (ISE)±0.5% FS (mono); 1% FS (div)
Accuracy (mV)±0.2 mV
Accuracy (pH)±0.002
Accuracy (RDO)0.1% ; ±0.2mg/L / ±0.2%
Accuracy (Salinity)±1% FS + 1 LSD
Accuracy (TDS)±1% FS +1 LSD
Alarm OutputCalibration Due, HI-LO limit (out of range reading)
Barometric Pressure CorrectionAutomatic correction with built-in sensor
Battery Type4 X 1.5 V AA Alkaline
Calibration Points (ISE)Up to 8
Calibration Points (pH)1 (Offset) to 6 points
Cell Constant0.010 to 10.000
DescriptionWaterproof CyberScan PCD 650 pH/ mV/ Ion/ Conductivity/ TDS/ Resistivity/ Salinity/ Dissolved Oxygen Meter with DAS software & multi probe holder. pH Buffer Options: USA, NIST, DIN, PWB, Custom
Display TypeMulti-data Backlit Dot Matrix, 7.2 cm (H) X 5.4 cm (W)
IP RatingIP67
Measurement ModespH, mV, Ion, Conductivity, TDS, Resistivity, Salinity, Dissolved Oxygen
Range (Conductivity)0 to 500 mS/cm
Range (mV)±2000.0 mV
Range (pH)-2 to 20
Range (Salinity)0 to 80 ppt
Range (TDS).. to 500.0 ppt
Resolution (ISE)2 or 3 digits
Resolution (pH)0.1/0.01/0.001
Temperature Range (Metric)-10.0°C to +110.0°C (Meter)
IncludesAuto-Calibration; Data-logging options with time/date indexed, GLP compliance; Simultaneous Display and measurement of 4 parameters; Cal-due alarm; pH slope/offset Display & Self-diagnostic
Memory500 Data Sets with Date/Time stamping & Security password protection
Power Supply9 V DC adapter, 500 mA
Pressure Resolution450 to 825 mmHg / 59.9 to 109.9 kP
Range (ISE)0.001 to 19900
Range (RDO)0 to 90.00 mg/L or ppm / 0 to 600%
Resolution (Conductivity)0.05% FS
Resolution (mV)0.1 mV
Resolution (RDO)0.01 mg/L or ppm
Resolution (Salinity)0.05% FS
Resolution (TDS)0.05% FS
Salinity FactorSalinity Correction = Automatic, Resolution & Accuracy = 0.0 to 50.0ppt & 0.1 ppt
Temperature Accuracy±0.5°C, ±0.9°F
Temperature CompensationAutomatic / Manual (0 to 100 °C); Temperature Coefficient = Linear & Pure
Temperature Range (English)14.0°F to 230.0°F (Meter)
Temperature Resolution0.1 °C/°F
Unit SizePK