▸
Facility Safety and Maintenance
▸
Emergency Eyewashes and Showers
▸
Eyewashes and Showers
▸
Eye Wash Station
Actual product may vary
Fisherbrand
Eye Wash Station
Keep the Eye Wash Station Refill on hand per the 1997 Approved Code of PracticeP L74 regulation for contingency supplies in case of emergency.
Have Questions?
Catalog number
FKS12980475
Price (SGD)
134.00
Each
Add to cart
Price (SGD)
134.00
Each
Add to cart
Wall mountable
Integrated mirror
Two 500mL bottles
Specifications
Includes
Two 500 mL Bottles and Mirror
Product Type
Eyewash Station