Stainless Steel Laboratory Funnel
Stainless Steel Laboratory Funnel
Actual product may vary
Fisher Catalogue
Stainless Steel Laboratory Funnel
Use with or without filter membranes for gravity filtration as well as simple funnel procedures. Nickel Electro™ Stainless Steel Laboratory Funnel is vented to allow complete, uninhibited flow of liquids or solids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopLength (Metric) StemHeight (Metric)
FKS12973621165 mm75 mm158 mm
FKS12963621103 mm55 mm115 mm
Catalog number FKS12973621
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric)Top:
165 mm
Length (Metric) Stem:
75 mm
Height (Metric):
158 mm
  • Durable stainless steel funnel, break-resistant, stain proof and easy to clean.
  • May be used with or without filter membranes for gravity filtration as well as simple funnel procedures.
  • Flute is vented to allow complete, uninhibited flow.
  • Can be used with liquids or solids.
  • The handle can also be used as a convenient hanging point when not in use.
  • Smooth finish on internal surfaces making funnel easy to clean or sterilise.
Specifications
Diameter (Metric)Top165 mm
MaterialStainless Steel
Height (Metric)158 mm
IncludesHandle
Length (Metric) Stem75 mm
Unit SizeEA