Eutech™ Refilling Reference Electrolyte (Double Junction pH electrode)
Product Image
Fisherbrand
Eutech™ Refilling Reference Electrolyte (Double Junction pH electrode)
Eutech™ offers Reference Standards and Solutions for use with Eutech™ pH electrodes
Have Questions?
Catalog NumberQuantity
FKS12973447480 mL
Catalog number FKS12973447
Price (SGD)
418.00
EA
Add to cart
Quantity:
480 mL
Price (SGD)
418.00
EA
Add to cart
The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers, calibration, and reference solutions for use with pH meters and electrodes.
Features:
  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
Chemical Name or MaterialRefilling Reference Electrolyte
DescriptionSaturated Potassium Chloride for Double Junction pH Electrodes
Solution TypeCalibration Solution
Industry TypeLaboratory Use
Quantity480 mL
Unit SizeEA