Midi Centrifuge
Midi Centrifuge
Actual product may vary
Fisherbrand
Midi Centrifuge
Achieve max speed with minimum space. Fisherbrand™ Midi Centrifuge is ideal for protocols calling for fast spins with a higher RCF requirement. Ideal for micro-filter cell separations, PCR, clinical applications and HPLC protocols. Includes two rotors: standard 12-place rotor with lid and eight-place strip rotor for four 0.2mL tube strips or 32 single tubes.
Have Questions?
Catalog number FKS12972041
Price (SGD)
3,344.00
EA
Add to cart
Price (SGD)
3,344.00
EA
Add to cart
  • 5 Year Warranty
  • Compact footprint and brush-less motor for quiet operation and low vibration
  • Select and set the digital speed display in RCF or RPM
  • Sample imbalance and tilt safety cut-out
  • Safety interlock lid prevents opening while rotor is turning
  • Back lit display showing time, speed and operational information on the same screen
  • Quick spin button for short spin requirements
  • UV resistant plastic, increased useful life under harsh light conditions
  • Push-button lid release
  • Manual override allows user to open lid in case of power failure
Specifications
Capacity12 x 1.5/2 mL Tubes (Rotor 1), 8 places for 4 x 0.2 mL Tube Strips or 32 x 0.2 mL Single Tubes (Rotor 2)
Max. RCF9,800 x g
DisplayDigital
TypeMidi Centrifuge
IncludesRotors, 12 x 0.2mL and 12 x 0.5mL tube adapters, universal double insulated power adapter with four interchangeable plugs
Electrical Requirements100/240 V 50/60 Hz
Depth (Metric)171 mm
Maximum Speed Range500 to 12,500 rpm
Rotor IncludedYes
Dimensions (HWD)114 x 171 x 203 mm
For Use With (Application)For PCR, microfilter separation, clinical applications and HPLC protocols.
Unit SizeEA