PCR Chamber 825-PCR/Hepa/Exp with Hepa Filtration Multiple User
PCR Chamber 825-PCR/Hepa/Exp with Hepa Filtration Multiple User
Actual product may vary
Fisherbrand
PCR Chamber 825-PCR/Hepa/Exp with Hepa Filtration Multiple User
Have Questions?
Catalog number FKS12964480
Price (SGD)
30,015.00
EA
Add to cart
Price (SGD)
30,015.00
EA
Add to cart
acrylic construction removable front sheild 220-240V 50Hz clear 210L Plas Labs