Quickfit™ Borosilicate Glass Iodine Flask
Quickfit™ Borosilicate Glass Iodine Flask
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Iodine Flask
Complies with BS 2735
Have Questions?
Catalog NumberCapacity (Metric)Standard Taper Joint
FKS12959333500 mL24/29
Catalog number FKS12959333
Price (SGD)
679.00
PK
Add to cart
Capacity (Metric):
500 mL
Standard Taper Joint:
24/29
Price (SGD)
679.00
PK
Add to cart
Complies with BS 2735
  • Complies with BS 2735
  • For the determination of iodine
  • Flask has ground socket and cup shaped top
  • Stopper has extended handle which projects well above the liquid seal
Note: Stopper is not included; sold separately.
Specifications
Certifications/ComplianceBS 2735
DescriptionTop style: Cup shaped
Capacity (Metric)500 mL
MaterialBorosilicate Glass, Borosilicate Glass
Standard Taper Joint24/29
Unit SizePK