Steel Wire Gauze
Steel Wire Gauze
Actual product may vary
Fisherbrand
Steel Wire Gauze
Provides excellent support to beakers and other glassware during heating on ring stands and tripods. R&L Enterprises™ Steel Wire Gauze is a flat square manufactured from steel mesh.
Have Questions?
Catalog NumberDimensions (L x W)
FKS12958950125 x 125 mm
Catalog number FKS12958950
Price (SGD)
61.00
PK
Add to cart
Dimensions (L x W):
125 x 125 mm
Price (SGD)
61.00
PK
Add to cart
  • Non-fraying edges
  • Durable and long-lasting
  • Excellent heat distribution
Specifications
Ceramic CenterCeramic Center
Dimensions (L x W)125 x 125 mm
EdgesNon-Fraying
Quantity10 Pack
ShapeSquare
Length (Metric)125 mm
MaterialSteel
TypeWire Gauze
Width (Metric)125 mm
Unit SizePK