MBL™ Double Ended Borosilicate Glass Stirring Rod
MBL™ Double Ended Borosilicate Glass Stirring Rod
Actual product may vary
Fisherbrand
MBL™ Double Ended Borosilicate Glass Stirring Rod
Minimize scratching and increase safety with these glass stirring rods. MBL™ Borosilicate Double Ended Glass Stirring Rods are ideal for a wide range of stirring applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS12958934200 mm
FKS12461627300 mm
Catalog number FKS12958934
Price (SGD)
133.00
PK
Add to cart
Length (Metric):
200 mm
Price (SGD)
133.00
PK
Add to cart
  • Manufactured from borosilicate glass
  • With rounded button and paddle ends
  • Button end is useful for crushing clumped powder and solids
  • Paddle end helps to circulate sediment when stirring
Specifications
For Use With (Application)General stirring applications
IncludesRounded button and paddle ends
Diameter (Metric)6 mm
Length (Metric)200 mm
MaterialBorosilicate Glass
Quantity10/Pk.
Unit SizePK