Sharpsguard™ yellow 11.5 Multi-Purpose 9.27L Sharps Container with Pail Handle
Sharpsguard™ yellow 11.5 Multi-Purpose 9.27L Sharps Container with Pail Handle
Actual product may vary
Fisherbrand
Sharpsguard™ yellow 11.5 Multi-Purpose 9.27L Sharps Container with Pail Handle
Features a robust pail handle for easy transportation and a wide aperture for easy insertion of larger items. Sharpsguard™ yellow 11.5 Multi-Purpose 9.27L Sharps Container with Pail Handle is for disposal of sharps that have been contaminated with medicinal products, but not cytotoxic or cytostatic.
Have Questions?
Catalog NumberCapacity (Metric)
FKS129534959.27 L
Catalog number FKS12953495
Price (SGD)
18.00
PK
Add to cart
Capacity (Metric):
9.27 L
Price (SGD)
18.00
PK
Add to cart
  • Container for disposal by incineration
  • Rigid polypropylene construction and wall thickness
  • Offers outstanding protection against penetrating sharps and leaking fluids
  • When container is full to the fill line it must be locked and disposed of safely
  • All have clear British Standard markings, user instructions and a visible fill line
Specifications
Capacity (Metric)9.27 L
Certifications/ComplianceBS7320:1990, UN3291, AFNOR, NFX 30-500
Closure TypeLid
ColorYellow
Height (Metric)242 mm
Length (Metric)280 mm
AutoclavableAutoclavable
Diameter (Metric) Opening275 mm
For Use With (Application)For easy transportation