Polystyrene Antistatic Weighing Boats
Polystyrene Antistatic Weighing Boats
Actual product may vary
Fisherbrand
Polystyrene Antistatic Weighing Boats
Weigh and pour samples with ease. The Fisherbrand™ Polystyrene Antistatic Weighing Boat folds easily and features a smooth surface and round corners for easy pouring. Choose the size and color that works best for your needs. Disposable and durable.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)ColorSizeShape
FKS1295286085 mLBlackMedium; 85 x 85 x 24 mmSquare
FKS1294286010 mLBlackSmall; 46 x 46 x 8 mmSquare
FKS12608513280 mLWhiteLarge; 140 x 140 x 22 mmDiamond
FKS1168030285 mLWhiteMedium; 85 x 85 x 24 mmDiamond
FKS1292285010 mLWhiteSmall; 46 x 46 x 8 mmSquare
Catalog number FKS12952860
Price (SGD)
174.00
PK
Add to cart
Capacity (Metric):
85 mL
Color:
Black
Size:
Medium; 85 x 85 x 24 mm
Shape:
Square
Price (SGD)
174.00
PK
Add to cart
  • Flexible boats feature smooth surfaces and rounded corners to provide accurate pour-outs with minimal sample loss
  • Select white or black for best color contrast
  • Antistatic surface to ensure sample is completely removed
  • Boats are shallow with a wide, flat bottom to resist tipping
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases
  • Temperature range -10 to 70°C
  • Available in three sizes and two colors, in packs of 500
  • All are FDA/food-grade compliant
Specifications
Bottom ShapeFlat
Capacity (Metric)85 mL
ColorBlack
DisposableSingle-use
MaterialPolystyrene
ShapeSquare
TypeAntistatic Weighing Boat
For Use With (Equipment)Microwave Moisture Analyzer
SizeMedium; 85 x 85 x 24 mm
Length (Metric)85 mm
Width (Metric)85 mm
Height (Metric)24 mm
Unit SizePK