Sharpsguard™ yellow 7 Multi-Purpose 5.8L Sharps Container with Pail Handle
Sharpsguard™ yellow 7 Multi-Purpose 5.8L Sharps Container with Pail Handle
Actual product may vary
Fisher Catalogue
Sharpsguard™ yellow 7 Multi-Purpose 5.8L Sharps Container with Pail Handle
Features a robust pail handle for easy transportation and a wide aperture for easy insertion of larger items. Sharpsguard™ yellow 7 Multi-Purpose 5.8L Sharps Container with Pail Handle is for disposal of sharps that have been contaminated with medicinal products, but not cytotoxic or cytostatic.
Have Questions?
Catalog NumberCapacity (Metric)
FKS129434955.8 L
Catalog number FKS12943495
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
5.8 L
  • Container for disposal by incineration
  • Rigid polypropylene construction and wall thickness
  • Offers outstanding protection against penetrating sharps and leaking fluids
  • When container is full to the fill line it must be locked and disposed of safely
  • All have clear British Standard markings, user instructions and a visible fill line
Specifications
Capacity (Metric)5.8 L
Certifications/ComplianceBS7320:1990, UN3291, AFNOR, NFX 30-500
Closure TypeLid
ColorYellow
Height (Metric)204 mm
Length (Metric)246 mm
AutoclavableAutoclavable
Diameter (Metric) Opening246 mm
IncludesClosure
For Use With (Application)For easy transportation