Microcentrifuge Tube, 1.5mL
Microcentrifuge Tube, 1.5mL
Actual product may vary
Fisherbrand
Microcentrifuge Tube, 1.5mL
Made from selected high-quality and biologically inert polypropylene. Leak-free caps stay closed during centrifugation with excellent visibility.
Have Questions?
Catalog NumberColor
FKS12942230Clear
Catalog number FKS12942230
Price (SGD)
100.00
PK
Add to cart
Color:
Clear
Price (SGD)
100.00
PK
Add to cart
  • Polypropylene graduated microcentrifuge tube 
  • Snap top cap
  • With write on label 
Specifications
Cap StyleSnap Cap
Closure ColorClear
Closure MaterialPolypropylene
Capacity (Metric)1.5 mL