Profile 3000 Nitrile Examination Gloves
Profile 3000 Nitrile Examination Gloves
Actual product may vary
Fisherbrand
Profile 3000 Nitrile Examination Gloves
Combine comfort and chemical resistance with Nitritex™ Profile 3000 nitrile exam gloves. Ambidextrous, latex-free glove offers chemical and puncture resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
FKS12940825Large
FKS12920825Small
Catalog number FKS12940825
Price (SGD)
-
Size:
Large
  • Nitrile is resistant to a wide range of chemicals
  • Excellent puncture resistance
  • Beaded cuff provides easier donning and stability
  • Textured surface for increased grip
  • Powder-free and latex-free to eliminate Type I latex allergies
  • Wrist-length gloves measure 240mm/9.5 in.
  • Easy-access 100-glove dispenser box
Specifications
Certifications/ComplianceISO 9001:2008, 13485:2003 and ISO 2859
ClassificationClass I Medical Device
ColorWhite
DisposableSingle-use
HandAmbidextrous
Latex FreeYes
MaterialNitrile
PowderedPowder Free
Product TypeNitrile Examination Glove
SizeLarge
SterilityNon-sterile
For Use With (Application)Premium examinations
Glove Cuff StyleBeaded/Rolled
Inspected for Medical UseYes
ModelProfile 3000
Packaging TypeBoxed
Quantity100/Pk.
SeriesProfile 3000
Length (Metric)24 cm
Unit SizePK