HM Series Heating Mantles
HM Series Heating Mantles
Actual product may vary
Fisher Catalogue
HM Series Heating Mantles
The HM series are a range of controlled heating mantles, available in a range of sizes to accomodate round bottom flasks from 50 to 1000 mL, constructed with a robust aluminum casing, making them tough and easy to clean.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Dimensions (D x W x H) Interior
FKS129401611000 mL220 x 220 x 170mm
FKS12930161500 mL170 x 220 x 220mm
Catalog number FKS12940161
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Dimensions (D x W x H) Interior:
220 x 220 x 170mm
  • Controlled up to 450°C
  • Range of sizes available
  • Integrated clamp rod fitting to enable secure fitting of a standard clamp rod
  • Non-slip feet are also fitted as standard
  • For added safety, all units feature an emergency cut out which will automatically trigger in the event of a spillage
Specifications
Capacity (Metric)1000 mL
Depth (Metric)220 mm
Dimensions (D x W x H) Interior220 x 220 x 170mm
Height (Metric)170 mm
Voltage230 V
Electrical Requirements230V, 50/60Hz, 380w
ModelHM1000C
MaterialAluminum
ShapeRound
Wattage380w
Width (Metric)220mm
Product TypeHeating Mantle
Temperature (Metric)450°C
For Use With (Application)Emergency cut out which will automatically trigger in the event of a spillage