Eutech™ Conductivity Calibration Sachets
Product Image
Fisherbrand
Eutech™ Conductivity Calibration Sachets
Get quick and accurate conductivity measurements in the field, laboratories, and schools.
Have Questions?
Catalog NumberConductivity
FKS1293343710 μS/cm
Catalog number FKS12933437
Price (SGD)
315.00
PK
Add to cart
Conductivity:
10 μS/cm
Price (SGD)
315.00
PK
Add to cart
Get quick, accurate, and convenient conductivity measurements with NIST_traceable Eutech™ Conductivity Calibration Sachets. Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed.
Features:
  • Conductivity Standards Sachet -20 ml
  • NIST Traceable
Specifications
Chemical Name or MaterialKCl Conductivity Sachets
Conductivity10 μS/cm
Solution TypeCalibration Solution
TypeConductivity Standard
Quantity20 x 20 mL
Unit SizePK