TST™ Control™ Self Adhesive Emulating Indicator Strip
TST™ Control™ Self Adhesive Emulating Indicator Strip
Actual product may vary
Fisherbrand
TST™ Control™ Self Adhesive Emulating Indicator Strip
Place the TST™ Control™ Self Adhesive Emulating Indicator Strip inside trays, packs or pouches when autoclaving so the indicator will confirm that good quality steam has penetrated to the point of placement.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature (Metric)Sterilization Time
FKS12926996121°C to 134°C5.3 and 15 min.
FKS12906996121°C to 134°C5.3 min.
Catalog number FKS12926996
Price (SGD)
297.00
PK
Add to cart
Temperature (Metric):
121°C to 134°C
Sterilization Time:
5.3 and 15 min.
Price (SGD)
297.00
PK
Add to cart
  • ISO 11140 part 1, class 6 classification
  • Paper
  • Steam sterilization
Specifications
AutoclavableAutoclavable
Certifications/ComplianceISO 11140 part 1, class 6
For Use With (Application)For sterilization
For Use With (Equipment)Autoclave
Product TypeEmulating Indicator Strip
Temperature (Metric)121°C to 134°C
FeaturesWhen placed inside trays, packs or pouches, a TST Control Emulating Indicator will confirm that good quality steam has penetrated to the point of placement
MaterialPaper
Quantity100 Pk.
Sterilization IndicatorYes
Sterilization Time5.3 and 15 min.
Sterilization MethodSteam
Unit SizePK