Stuart™ HM100C Model Heating Mantle
Stuart™ HM100C Model Heating Mantle
Actual product may vary
Fisherbrand
Stuart™ HM100C Model Heating Mantle
Have Questions?
Catalog number FKS12910161
Price (SGD)
3,483.00
EA
Add to cart
Price (SGD)
3,483.00
EA
Add to cart
Specifications
Capacity (Metric)100 mL
Depth (Metric)175 mm
Dimensions (D x W x H) Interior175 x 175 x 170mm
Height (Metric)150 mm
Voltage230 V
Electrical Requirements230V, 50/60Hz, 100w
ModelHM100C
MaterialAluminum
ShapeRound
Wattage100w
Width (Metric)175mm
Product TypeHeating Mantle
Temperature (Metric)450°C
For Use With (Application)Emergency cut out which will automatically trigger in the event of a spillage