Sharpsguard™ yellow 1 0.84L Multi-Purpose Sharps Container with Push-Down Lid
Sharpsguard™ yellow 1 0.84L Multi-Purpose Sharps Container with Push-Down Lid
Actual product may vary
Fisher Catalogue
Sharpsguard™ yellow 1 0.84L Multi-Purpose Sharps Container with Push-Down Lid
Ideal for phlebotomist usage with a push-down, one-piece lid which incorporates a vacutainer remover. Sharpsguard™ yellow 1 0.84L Multi-Purpose Sharps Container with Push-Down Lid is for disposal of sharps that have been contaminated with medicinal products, but not cytotoxic or cytostatic.
Have Questions?
Catalog NumberCapacity (Metric)
FKS129034950.84 L
Catalog number FKS12903495
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
0.84 L
  • Supplied with lid assembled
  • Will fit securely into Sharpsguard tray range
  • Container for disposal by incineration or other means
  • Rigid polypropylene construction and wall thickness
  • Offers outstanding protection against penetrating sharps and leaking fluids
  • When container is full to the fill line it must be locked and disposed of safely
  • All have clear British Standard markings, user instructions and a visible fill line
Specifications
Capacity (Metric)0.84 L
Certifications/ComplianceBS EN ISO 23907.
Closure TypeLid
ColorYellow
Height (Metric)127 mm
Length (Metric)205 mm
Opening Size (Metric)70 x 32 mm (L x W)
AutoclavableAutoclavable
For Use With (Application)Ideal for phlebotomists