Vacuum Pump Oil
Vacuum Pump Oil
Actual product may vary
Fisherbrand
Vacuum Pump Oil
One and two-stage rotary vane pump. Fisherbrand™ Vacuum Pump Oil designed for use with the range of Fisherbrand two-stage rotary vane pumps and with most other one and two-stage rotary vane pump.
Have Questions?
Catalog NumberVolume (Metric)
FKS129011511 L
Catalog number FKS12901151
Price (SGD)
125.00
EA
Add to cart
Volume (Metric):
1 L
Price (SGD)
125.00
EA
Add to cart
One and two-stage rotary vane pump. Fisherbrand™ Vacuum Pump Oil designed for use with the range of Fisherbrand two-stage rotary vane pumps and with most other one and two-stage rotary vane pump.
Generic mineral oil for use in most one and two stage rotary vane pumps.
Specifications
Product TypeVacuum Pump Oil
For Use With (Application)For General Use in Rotary Vane Pumps
For Use With (Equipment)Rotary Vane Pumps
Pressure10-5mabr vapour pressure
Viscosity118 cSt viscosity at 40°C
Volume (English)33.81 oz.
Volume (Metric)1 L
Unit SizeEA