Distel™ Surface Disinfectant
Distel™ Surface Disinfectant
Actual product may vary
Fisherbrand
Distel™ Surface Disinfectant
Tristel™ Distel™ Surface Disinfectant is an all-purpose, non-alcohol based formula for use in cleaning and disinfecting laboratory surfaces and equipment.
Have Questions?
Catalog NumberVolume (Metric)CompositionContainerType
FKS128993575000 mLDistel TM309
Catalog number FKS12899357
Price (SGD)
-
Volume (Metric):
5000 mL
Composition:
Distel TM309
  • Classified as a biocide under the EU Biocides Regulation
  • Tested and approved as bactericidal, fungicidal, virucidal and tuberculocidal
  • Effective in the presence of blood borne diseases (HIV/Hepatitis B) and also destroys DNA and RNA
  • Recommended for use at a dilution of 1:100 for all normal laboratory requirements
  • Suitable for use on most surfaces including glass, metal, plastic, rubber and fabric
Specifications
Product TypeSurface Disinfectant
FormConcentrate
CompositionDistel TM309
Dilution0.11111111111
For Use With (Application)Laboratory equipment, Benches and shelves, Fridges, pipettors, Water baths, Incubators, Centrifuges, Discard jars, Non-critical devices
Shelf Life3 Years
Volume (Metric)5000 mL
Unit SizePK