Sodium Arsenite, 98+%, Extra Pure, SLR, Fisher Chemical
Actual product may vary
Sodium Arsenite, 98+%, Extra Pure, SLR, Fisher Chemical
Sodium Arsenite, 98+%, Extra Pure, SLR, Fisher Chemical
Fisherbrand
Sodium Arsenite, 98+%, Extra Pure, SLR, Fisher Chemical
Have Questions?
Catalog NumberQuantityPackaging
FKS12897692100 gPlastic powder jar
Catalog number FKS12897692
Price (SGD)
-
Quantity:
100 g
Packaging:
Plastic powder jar
100g, Plastic powder jar
Specifications
Melting Point615°C
CAS7784-46-5
ColorWhite
pH12
Physical FormSolid
Quantity100 g
Molecular FormulaAsNaO2
MDL Number3472
Synonymsodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c
InChI KeyPTLRDCMBXHILCL-UHFFFAOYSA-M
SMILES[O-][As]=O.[Na+]
IUPAC Namesodium;oxoarsinite
Molecular Weight (g/mol)129.909
PubChem CID443495
ChEBICHEBI:29678
Formula Weight129.91g/mol
PackagingPlastic powder jar
Unit SizeEach