Mini Chromato-Vue™ Viewing Cabinets
Actual product may vary
Fisherbrand
Mini Chromato-Vue™ Viewing Cabinets
Provides a compact darkroom environment for viewing materials. UVP™ Mini Chromato-Vue™ Viewing Cabinets include a contrast control filter that absorbs UV energy to protect the eyes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)Model
FKS12893828Mini Viewing Cabinet4W EL LampsC-10E4
FKS11778201Mini Viewing CabinetCompact/Handheld LampsC-10
Catalog number FKS12893828
Price (SGD)
1,567.00
Each
Product Type:
Mini Viewing Cabinet
For Use With (Equipment):
4W EL Lamps
Model:
C-10E4
Price (SGD)
1,567.00
Each
  • Each cabinet is designed to be used with specific UVP lamp models, which fit snugly on top of the cabinet
  • Mini viewing cabinets are constructed with a metal base and plastic top
  • The bottom panel of the cabinet is removable
  • A contrast control filter absorbs UV energy to protect the eyes
  • Soft rubber viewport provides a comfortable viewing area with adequate space for eyeglasses
  • Lightweight access curtain (Cat. No. UVP72-0001-01) blocks out external light from the cabinet interior
C-10P: Acommodates rechargeable lamps
C-10: To be used with compact and handheld UV lamps
C-10E4: Designed for use with the 4-watt EL Series lamps
C-10E6: Designed for use with the 6-watt EL Series lamps
C-15G: Combines the C-10 cabinet with the UVGL-58 multi-band shortwave/longwave handheld Lamp to provide a self-contained illumination system
Applications:
Mineralogy, Lab fluorescence, Sterilization, Field investigation
Specifications
Depth (Metric)229mm
For Use With (Equipment)4W EL Lamps
Height (Metric)305mm
Width (Metric)267mm
ModelC-10E4
For Use With (Application)for viewing materials
Product TypeMini Viewing Cabinet
Unit SizeEach