Stomacher™ Model 400 Sterile Bags
Product Image
Fisherbrand
Stomacher™ Model 400 Sterile Bags
The Stomacher™ Bag range has been designed to fulfill all applications from sample blending, straining and storage, including bags designed for extensive blending and small samples for life science applications.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1288940580 to 400 mL
Catalog number FKS12889405
Price (SGD)
-
Capacity (Metric):
80 to 400 mL
The Stomacher™ Bag range has been designed to fulfill all applications from sample blending, straining and storage, including bags designed for extensive blending and small samples for life science applications.
  • Manufactured from high quality food grade virgin LDPE granule extruded to 65μm film thickness
  • Designed to withstand extensive blending without tears or perforations
  • Provides a low-cost option for sample preparation with no risk of cross-contamination between samples
  • All bags are irradiated sterile; certificate of irradiation is available upon request
  • Designed for blended homogenization of diluent samples for most applications
Specifications
DescriptionThickness of 65 microns
Capacity (Metric)80 to 400 mL
For Use With (Application)Blended homogenization of diluent samples
For Use With (Equipment)Stomacher 400
MaterialLDPE
Outer PackagingSupplied as 20 sachets of 50 bags
Quantity1000 (sachets of 50)
Dimensions (L x W)177 x 305 mm
Unit SizePK