Check Order Status
Polyethylene Pasteur Pipets
Actual product may vary
Fisherbrand
Polyethylene Pasteur Pipets
Use these pipettes for a variety of liquid handling applications. Medline™ Polyethylene Pasteur Pipets are made of transparent, inert and low intensity polyethylene.
Have Questions?
Catalog Number
Volume (Metric)
FKS12887605
3 mL
Catalog number
FKS12887605
Price (SGD)
326.00
PK
Add to cart
Volume (Metric):
3 mL
Price (SGD)
326.00
PK
Add to cart
Nonsterile
Graduated each 0.5mL up to 5mL
Volumetrical tolerance: ±10%
Specifications
Diameter (Metric) Outer
7.8 mm
Material
Polyethylene
Sterility
Non-sterile
Quantity
500 Pack
Top Style
Bulb Shaped
Packaging Type
Sealed
Color
Transparent
Volume (Metric)
3 mL
Length (Metric)
154 mm
Type
Pasteur Pipet
Disposable
Single-use