Check Order Status
Sterilin™ Silicone Tubing
Actual product may vary
Fisher Catalogue
Sterilin™ Silicone Tubing
Perform various applications with high quality, silicone tubing made from nontoxic material.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Inner
Diameter (Metric) Outer
Thickness (Metric)
FKS12883003
6 mm
14 mm
4 mm
FKS12853013
8 mm
12 mm
2 mm
2 Options
Catalog number
FKS12883003
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric) Inner:
6 mm
Diameter (Metric) Outer:
14 mm
Thickness (Metric):
4 mm
Perform various applications with high-quality silicone tubing made from nontoxic material.
Ideal for use with peristaltic pumps
Can be autoclaved or dry heat sterilized
15-meter length
Manufactured in ISO Class 7 clean room
Specifications
Description
Silicone tubing
Autoclavable
Autoclavable
Diameter (Metric) Inner
6 mm
Material
Silicone
Diameter (Metric) Outer
14 mm
Overall Length (English)
49 ft.
Overall Length (Metric)
15 m
Thickness (Metric)
4 mm
Color
Translucent
Flexibility
Very Flexible
Product Type
Flexible Tubing