Test Tube Brushes with Nylon Head
Test Tube Brushes with Nylon Head
Actual product may vary
Fisherbrand
Test Tube Brushes with Nylon Head
Features fan shaped end. Pinnacle Brush™ Test Tube Brushes with Nylon Head thoroughly cleans test tubes.
Have Questions?
Catalog NumberBristle Material
FKS12881954Nylon
Catalog number FKS12881954
Price (SGD)
45.00
PK
Add to cart
Bristle Material:
Nylon
Price (SGD)
45.00
PK
Add to cart
  • With fan-shaped end
  • Galvanised steel wire handle
  • Nylon fill
Specifications
Product TypeTest Tube Brush
Bristle MaterialNylon
Handle MaterialMetal
Diameter (Metric)11.5 mm
For Use With (Equipment)Suitable for use with 10 to 12mm test tubes
Length (Metric)19 cm
Length (Metric) Brush6 cm
Length (Metric) Handle13 cm
Quantity10 Pk.
SizeBrush Circumference: 5 cm
Unit SizePK