Men's Fitted Laboratory Coat
Men's Fitted Laboratory Coat
Actual product may vary
Fisher Catalogue
Men's Fitted Laboratory Coat
Logistik Unicorp™ Men's Fitted Laboratory Coat is hardwearing, practical, and designed for comfort.
Have Questions?
Catalog NumberColorSize (Metric) Chest
FKS12881121Spruce Green100 cm
Catalog number FKS12881121
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Spruce Green
Size (Metric) Chest:
100 cm
  • Manufactured in hardwearing polycotton material making it a practical, comfortable garment with all the durability that you would expect from a coat of this quality
  • Concealed stud front
  • One breast pocket
  • Two lower patch pockets
  • Center back vent for ease of movement
  • Material 65% polyester/35% cotton 245 g/m2
  • Not CE marked
Specifications
Product TypeGeneral Purpose Lab Coat
GenderMen's
MaterialPolycotton
ColorSpruce Green
DisposableReusable
SterilityNon-sterile
No. of Pockets3
Size (Metric) Chest100 cm
For Use With (Application)General Laboratory
IncludesOne breast pocket, two lower patch pockets, center back vent
Weight (Metric)245 g/m2
Unit SizeEA