GHS Health Hazard Labels, 20 x 20mm
GHS Health Hazard Labels, 20 x 20mm
Actual product may vary
Fisherbrand
GHS Health Hazard Labels, 20 x 20mm
Laminated and produced on a polypropylene material. Limpet Labels GHS Health Hazard Labels conform with B.S 5609:1986 which is perfect for transport by road, rail, air, and sea.
Have Questions?
Catalog NumberMaterial
FKS12879758Polypropylene
Catalog number FKS12879758
Price (SGD)
-
Material:
Polypropylene
  • Hazard warning diamonds fully comply with the GHS, CLP, RID IMDG/IMO, ADN, & ICAO/IATA requirements
  • Excellent weather, chemical, and abrasion resistance
  • Marine grade adhesive
  • 1000 labels per roll
Specifications
Certifications/ComplianceGHS, CLP, RID IMDG/IMO, ADN, & ICAO/IATA requirements
FinishLaminated
MaterialPolypropylene
Quantity1000/Roll
Resistant ToWeather, chemicals, abrasion
DescriptionPP (Polypropylene)
Mounting TypeSelf-adhesive
Height (Metric)20 mm
Width (Metric)20 mm
Unit SizePK