Tabletop spill tray, 11.3L capacity
Tabletop spill tray, 11.3L capacity
Actual product may vary
Fisherbrand
Tabletop spill tray, 11.3L capacity
Have Questions?
Catalog number FKS12878909
Price (SGD)
742.00
EA
Add to cart
Price (SGD)
742.00
EA
Add to cart
Catch spills or leaks from cracked containers, bottles, etc. Built-in pouring spout for quick and easy sump drainage. Chemical-resistant polyethylene will not corrode. Translucent sidewalls make it easy to spot leaks.
  • Capacity: 11.3L
  • Dimensions (L x W): 530mm x 430mm