IPA Resistant LabMarker
IPA Resistant LabMarker
Actual product may vary
Fisherbrand
IPA Resistant LabMarker
A fine tipped permanent marker with alcohol-resistant ink that marks on cold or wet surfaces, plastic bags and disposable lab-ware.
Have Questions?
Catalog NumberColor
FKS12877053Black
Catalog number FKS12877053
Price (SGD)
262.00
PK
Add to cart
Color:
Black
Price (SGD)
262.00
PK
Add to cart
With permanent, quick drying black ink that is resistant to IPA
The Micronova IPA Resistant LabMarker features permanent, quick drying, alcohol-resistant ink that marks on a range of surfaces and will not smear on reusable glass, metal or porcelain, but can be scrubbed off. Available gamma irradiated for aseptic areas.
  • Fine-tip, black permanent marker
  • Quick drying, alcohol-resistant ink
  • Marks on cold or wet surfaces, plastic bags, and disposable lab-ware
  • Smear-resistant on glass, metal and porcelain, but can be scrubbed off
  • Available gamma irradiated for aseptic areas
Specifications
Product TypeCleanroom Pen
Tip StyleFine
ColorBlack
AutoclavableNot Autoclavable
SterilityNon-sterile
Material (Barrel)High-Density Polypropylene
DisposableReusable
For Use With (Application)Cleanroom and Aseptic Environment Documentation and Labeling
No. per Pack10
QuantityEa.
Capacity (Metric)2 mL
Length (English)5.25 in.
Unit SizePK