Medium Wave UV Tube
Medium Wave UV Tube
Actual product may vary
Fisherbrand
Medium Wave UV Tube
Medium Wave UV Tube for use in UV transilluminators.
Have Questions?
Catalog number FKS12873998
Price (SGD)
178.00
PK
Add to cart
Price (SGD)
178.00
PK
Add to cart
  • For use with all GX and GE model transilluminators
  • 302 nm wavelength
  • Medium wave
Specifications
Base Style2-pin
Gas FillNeon-mercury
Light SourceUV
Lighting Life3000 to 9000 hr.
Voltage100/115/220/230 V
Wattage50 W
Wavelength302 nm
Weight (English)<1 lb.
Weight (Metric)<0.5 kg
For Use With (Equipment)All GX/GE model transilluminators
Product TypeMidwave UV Tube
Unit SizePK