XX-15 Series UV Bench Lamp, 230V
XX-15 Series UV Bench Lamp, 230V
Actual product may vary
XX-15 Series UV Bench Lamp, 230V
XX-15 Series UV Bench Lamp, 230V
XX-15 Series UV Bench Lamp, 230V
Fisherbrand
XX-15 Series UV Bench Lamp, 230V
Bench lamps that offer a choice between longwave, shortwave or midrange wavelengths for use in sterilization/sanitation and fluorescence/inspection applications. UVP XX-15 Series UV Bench Lamp, 230V are 15 watt fixtures designed for use in Europe.
Have Questions?
Catalog NumberModelWavelengthLight Source
FKS12872969XX-15L365 nmUV-A
Catalog number FKS12872969
Price (SGD)
3,100.00
EA
Add to cart
Model:
XX-15L
Wavelength:
365 nm
Light Source:
UV-A
Price (SGD)
3,100.00
EA
Add to cart
  • Various UV tube wavelengths are available for different application needs
  • Lamps include holes on each end of the fixture for hanging or mounting the lamp
  • Manufactured with a durable aluminum housing and powder paint scratch resistant finish
  • Shortwave lamps can be used in an upward facing position for purifying air or in a downward position for decontamination of work surfaces
  • 254nm UV can kill up to 99% of organisms from bacteria, yeast, mold spores, germs and viruses
  • Shortwave UV offers a reliable and effective tool for decontamination applications in hospitals, laboratories, pharmaceutical companies and clinics
  • UV intensity can be increased by bringing the lamp closer to object being sterilized
  • Interchangeable UV tubes (choose 15 watt tubes for use with XX-15 lamps)
  • 15 Watt lamps: Size: 19.75L x 6W x 4.25 in.H (502 x 152 x 108mm), Weight: 5 lb (2.3kg)
Specifications
Light SourceUV-A
Voltage230 V
Wattage15 W
Wavelength365 nm
Dimensions (L x W x H)502 x 152 x 108 mm
Electrical Requirements15 W, 230 V, 60 Hz
For Use With (Application)Hospitals, laboratories, pharmaceutical companies and clinics
Height (Metric)108 mm
Hertz60 Hz
Length (Metric)502 mm
MaterialAluminum
ModelXX-15L
Packaging Quantity1 Ea.
Product LineXX-15 Series
Width (Metric)152 mm
Product TypeUV Bench Lamp
Unit SizeEA