Check Order Status
Norell™ Borosilicate Glass J Young NMR Valves
Actual product may vary
Fisherbrand
Norell™ Borosilicate Glass J Young NMR Valves
Have Questions?
Catalog Number
Length (Metric)
Field Strength
FKS12872306
178 mm
400 MHz
Catalog number
FKS12872306
Price (SGD)
491.00
EA
Add to cart
Length (Metric):
178 mm
Field Strength:
400 MHz
Price (SGD)
491.00
EA
Add to cart
Specifications
Length (Metric)
178 mm
Material
Borosilicate Glass
Product Type
J Young NMR Valve
Field Strength
400 MHz
For Use With (Application)
400MHz tube
Includes
Female joint
Quantity
1 Each