Screw Cap Container
Screw Cap Container
Actual product may vary
Fisherbrand
Screw Cap Container
Spoon screw cap makes this container ideal for collection and transportation of specimens. Kartell™ Screw Cap Container is available in transparent PS (Polystyrene) and comes completely assembled and ready to use.
Have Questions?
Catalog NumberSterility
FKS12870923Sterile
Catalog number FKS12870923
Price (SGD)
626.00
EA
Add to cart
Sterility:
Sterile
Price (SGD)
626.00
EA
Add to cart
  • Includes screw cap
  • Includes spoon
  • Capacity of 60mL
  • 65mm in height
  • Made of transparent PS
Specifications
MaterialPolystyrene
SterilitySterile
Capacity (Metric)60 mL
Certifications/ComplianceCE/IVD
Closure ColorRed
Closure MaterialPolyethylene
Closure TypeScrew Cap
Diameter (Metric)39 mm
Height (Metric)65 mm
IncludesWhite spoon
Unit SizeEA