Hazard Warning Label 'Irritant' Self-Adhesive GHS and CLP Compliant 20x20mm
Product Image
Fisherbrand
Hazard Warning Label 'Irritant' Self-Adhesive GHS and CLP Compliant 20x20mm
Have Questions?
Catalog number FKS12869758
Price (SGD)
-
All our Hazard Warning Diamonds are laminated and produced on a polypropylene material. The base material we use conforms with B.S 5609:1986 which is perfect for transport by Road, Rail, Air and Sea. Our Hazard Warning diamonds fully comply with the GHS, CLP, RID IMDG/IMO, ADN and ICAO/IATA requirements.
They have the following main properties:
• Excellent Weather Resistance.
• Excellent Chemical Resistance.
• Excellent Abrasion Resistance.
• Marine Grade Adhesive.