Phadebas™ Amylase Test Tablets (50)
Phadebas™ Amylase Test Tablets (50)
Actual product may vary
Fisher Catalogue
Phadebas™ Amylase Test Tablets (50)
Determine Α-amylase activitiy for applications in fields including the food industry, biotech, and detergents. Fisherbrand™ Phadebas™ Amylase Test Tablets come with instructions and a batch standard curve.
Have Questions?
Catalog NumberQuantity
FKS1286598350 Tablets
Catalog number FKS12865983
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 Tablets
  • For a wide variety of applications in fields including the food industry, biotech, and detergents.
  • This test has been on the market for over 35 years and is well documented in the scientific literature.
  • Includes instructions for use in English, French, Spanish, and German.
  • Includes a batch standard curve for quantitative assessment.
Specifications
For Use With (Application)Determining Α-amylase activity
Quantity50 Tablets
Unit SizeEA