Compact and Handheld UV Lamp Replacement Tubes
Compact and Handheld UV Lamp Replacement Tubes
Actual product may vary
Fisherbrand
Compact and Handheld UV Lamp Replacement Tubes
For use with 4w and 6w UV lamps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)VoltageWattageWavelength
FKS12862979EL-series UV lamp, 3UV™ lamp, UVSL-26P rechargeable UV lamp, handheld UV lamp and mid size UV viewing cabinets230 V6 W365 nm
FISUVP34001301Compact 4-Watt UV Lamp115 V6 W254 nm
FKS11902041Compact 4-Watt UV Lamp115 V6 W254 nm
FISUVP34003401EL-series UV lamp, 3UV™ lamp, UVSL-26P rechargeable UV lamp, handheld UV lamp and mid size UV viewing cabinets230 V6 W365 nm
Catalog number FKS12862979
Price (SGD)
94.00
EA
Add to cart
For Use With (Equipment):
EL-series UV lamp, 3UV™ lamp, UVSL-26P rechargeable UV lamp, handheld UV lamp and mid size UV viewing cabinets
Voltage:
230 V
Wattage:
6 W
Wavelength:
365 nm
Price (SGD)
94.00
EA
Add to cart
  • For use with 4w and 6w UV lamps
Specifications
Light SourceUV-A
Voltage230 V
Wattage6 W
Wavelength365 nm
Diameter (English)0.62 in.
Diameter (Metric)15.8 mm
For Use With (Equipment)EL-series UV lamp, 3UV™ lamp, UVSL-26P rechargeable UV lamp, handheld UV lamp and mid size UV viewing cabinets
Length (English)9 in.
Length (Metric)228.6 mm
Packaging Quantity1 Ea.
Product TypeUV Lamp Tube
Unit SizeEA