Check Order Status
Bunsen Safety Burner for LP Gas
Actual product may vary
Fisherbrand
Bunsen Safety Burner for LP Gas
Supplied with a flame failure device that shuts off the gas supply to the burner within approximately 20 seconds of the flame being extinguished.
Have Questions?
Catalog Number
Gas Type
FKS12862144
Natural Gas, Methane
Catalog number
FKS12862144
Price (SGD)
298.00
EA
Add to cart
Gas Type:
Natural Gas, Methane
Price (SGD)
298.00
EA
Add to cart
Safety features all contained within the burner
For use with natural gas/methane
12.7mm diameter opening
Specifications
Gas Type
Natural Gas, Methane
No. per Pack
1 Ea.