Howie-Style Unisex White Laboratory Coats with Knitted Cuffs
Actual product may vary
Fisher Catalogue
Howie-Style Unisex White Laboratory Coats with Knitted Cuffs
Includes two lower pockets. Logistik Unicorp™ Howie-Style Unisex White Laboratory Coats with Knitted Cuffs feature a wrapover style stud fastened front for extra frontal protection.
Have Questions?
Change view
Catalog Number
Size
Length (Metric)
FKS12861388
X-Large
100 cm
FKS12841388
Medium
100 cm
FKS12851388
Small
100 cm
FKS12348647
Large
100 cm
4 Options
Catalog number FKS12861388
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Size:
X-Large
Length (Metric):
100 cm
Includes two lower pockets. Logistik Unicorp™ Howie-Style Unisex White Laboratory Coats with Knitted Cuffs feature a wrapover style stud fastened front for extra frontal protection.
Howie style laboratory coat
Knitted cuffs
White
Two lower pockets
Wrapover style stud fastened front
Polyester/cotton
Not CE marked
Specifications
Product TypeGeneral Purpose Lab Coat
SizeX-Large
GenderUnisex
MaterialCotton, Polyester
ColorWhite
Cuff StyleKnit
DisposableReusable
SterilityNon-sterile
StyleHowie
No. of Pockets2
Collar TypeRound
Size (Metric) Chest116.84 to 121.92 cm
IncludesWith two lower pockets and wrap over style stud fastened front