Faithful Spruce Green Color 65% Polyester/35% Cotton Lab Coats
Faithful Spruce Green Color 65% Polyester/35% Cotton Lab Coats
Actual product may vary
Fisher Catalogue
Faithful Spruce Green Color 65% Polyester/35% Cotton Lab Coats
Spruce green lab coat for general laboratory use
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize (English) ChestSize (Metric) Chest
FKS1286112136.2 in.92 cm
FKS1280113142.5 in.108 cm
FKS1282113145.7 in.116 cm
FKS1283113148.8 in.124 cm
FKS1284113152 in.132 cm
Catalog number FKS12861121
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size (English) Chest:
36.2 in.
Size (Metric) Chest:
92 cm
This 105 cm long men's coat is manufactured in hardwearing polycotton material making it a practical, comfortable garment with all the durability that you would expect from a coat of this quality. Concealed stud front. One breast pocket. Two lower patch pockets. Centre back vent for ease of movement. Material 65% polyester/35% cotton 245g/m. Not CE marked.
Specifications
Product TypeGeneral Purpose Lab Coat
SizeX-Small
GenderUnisex
DisposableReusable
SterilityNon-sterile
Size (English) Chest36.2 in.
Size (Metric) Chest92 cm
For Use With (Application)General Laboratory
IncludesOne breast pocket, two lower patch pockets, centre back vent
Weight (Metric)245 g/m2
Unit SizePK